Learn More
Synthware™ Replacement Double Oblique Bore Stopcock
Specifications
Specifications
| Type | Double Oblique Stopcock |
| Plug Bore | 4 mm |
| Material | Borosilicate Glass |
| For Use With (Application) | Organic Synthesis |
| For Use With (Equipment) | Double Oblique Tap |
| Plug Material | PTFE |