Learn More
Contec Replacement Collars
Catalog No. 17720592
Change view
Click to view available options
Includes:
1 Collar Pole
3 Collar Pole
5 Collar Pole
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
17720592 1 Collar Pole
17720591 3 Collar Pole
17720593 5 Collar Pole
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17720592 Supplier Contec Supplier No. L3270114
Interchangeable mop handles, available in carbon fiber, anodised aluminum, stainless steel, or fiberglass.
  • Compatible with all QuickConnect™ frames
  • Multiple lengths available
  • Fully autoclavable QuickConnect™ mop handles and interchangeable mop head frames work together for maximum flexibility when cleaning in critical environments.
  • Allows investment in one handle with various interchangeable heads required for specific applications.
  • Handles are available in several lengths and styles.
  • Telescoping handles are available in lightweight carbon fiber, anodised aluminum, durable stainless steel, or fiberglass.
  • Choose from a variety of lengths to suit different cleaning needs.
Specifications
Product Type Cleanroom Mop Handle
Sterility Non-sterile
Autoclavable Autoclavable
Includes 1 Collar Pole
Color-coded No
Material Carbon Fiber