Learn More
Parker Hannifin Corporation Parker Hannifin Replacement Catalyst Tower, Hydrocarbon Removal, For Parker HPZA-7000 Generator
Catalog No. NC9040414
Encompass
The Parker Hannifin replacement catalyst tower purifies compressed air for the Parker HPZA-7000 Zero Air Generator, ensuring ultra-high purity output with less than or equal to 0.05 ppm total hydrocarbons. This component enhances analytical accuracy, reduces detector cleaning needs, and maintains silent generator operation. Its annual replacement requirement ensures optimal performance, making it an essential asset for applications such as Gas Chromatography.
Catalog No. NC9040414 Supplier Parker Hannifin Corporation Supplier No. 76820
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The Parker Hannifin replacement catalyst tower purifies compressed air for the Parker HPZA-7000 Zero Air Generator, ensuring ultra-high purity output with less than or equal to 0.05 ppm total hydrocarbons. This component enhances analytical accuracy, reduces detector cleaning needs, and maintains silent generator operation. Its annual replacement requirement ensures optimal performance, making it an essential asset for applications such as Gas Chromatography.