Learn More
Renewable Energy Kit
Catalog No. S98609
Change view
Click to view available options
For Use With (Application):
Demonstrates the workings of a complete clean energy system on a miniature scale
Catalog No. For Use With (Application)
S98609 Demonstrates the workings of a complete clean energy system on a miniature scale
1 options
Catalog No. S98609 Supplier Customer Channels Group (Ccg) Supplier No. FCJJ27
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes
Wind turbine kit, solar photovoltaic panel, electrolyzer, a PEM fuel cell, hydrogen storage system, all necessary parts, and a Renewable Energy Education CD
Requires
Scissors and water
Demonstrate the workings of a complete clean energy system on a miniature scale.
  • Combines electronics, chemistry, and physics with multiple experiments
  • Modular experiment set encourages users to learn the system step by step or configure the system in different ways
Renewable Energy Education CD
  • Renewable Energy Science Education Manual: Introduces renewable energy in seven chapters—Global Warming and Climate Change, Solar Technologies, Wind Technologies, Electrolyzers, Fuel Cells, Hydrogen Storage, and Basic Electronics
  • Experiment Manual: More than 20 experiments for middle to high school level sciences in several sections—Solar Energy, Wind Energy, Energy from Hydrogen (Fuel Cell), and “Ultra Cool« experiments
  • Flash Animations: Describe and show how a fuel cell works, different types of fuel cells, electrolysis, and various applications for fuel cells
  • Assembly Guides
  • Technical Support with videos
Specifications
For Use With (Application) Demonstrates the workings of a complete clean energy system on a miniature scale
Includes Wind turbine kit, solar photovoltaic panel, electrolyzer, a PEM fuel cell, hydrogen storage system, all necessary parts, and a renewable energy education CD
Product Type Renewable Energy Kit