Learn More
New England Biolab Ltd Remove-iT PNGase F 6750 units
Catalog No. NC0437310
Encompass
Remove-iT PNGase F is a tagged version of PNGase F, optimized for easy removal from a reaction. Remove-iT PNGase F is an amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides from N-linked glycoproteins.
Catalog No. NC0437310 Supplier New England Biolab Ltd Supplier No. P0706S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Remove-iT PNGase F is a tagged version of PNGase F, optimized for easy removal from a reaction. Remove-iT PNGase F is an amidase, which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides from N-linked glycoproteins.