missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Removal Tool
Description
- Removal tool for 25 mL adapter
- For SmartBlock 50 mL
Specifications
Specifications
| For Use With (Equipment) | 25 mL Adapter, SmartBlock 50 mL |
| Type | Removal Tool |