Learn More
GA International Removable Deep-Freeze Labels for Thermal-Transfer Barcode Printers 2"" x 1"" / 50.8mm x 25.4mm Rectangle Orange From -80C to +100C (-112F to +212F)
Catalog No. 5000572170
Encompass
Deep-freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (-20C, -40C, -80C) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.Waterproof and tearproof, these removable deep-freeze labels can also withstand temperatures ranging up to +100C, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.
Catalog No. 5000572170 Supplier GA International Supplier No. RMTT28C11OR
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Deep-freeze, removable barcode labels for vials, tubes, as well as polypropylene and cardboard boxes. The freezer labels have a strong, glove-friendly adhesive that allows them to adhere firmly to containers in (-20C, -40C, -80C) freezers and dry ice while still being easily removable at room temperature without leaving behind any residue.Waterproof and tearproof, these removable deep-freeze labels can also withstand temperatures ranging up to +100C, making them suitable for general lab use such as the identification of beakers, flasks, boxes, plastic and glass bottles that are stored in fridges and freezers.