missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Remote Dispensing System and Accessories for Dispensette™ Bottletop Dispensers

Catalog No. 13880710
Change view
Click to view available options
Product Type:
FEP Tubing
Remote Dispensing System
Length (Metric):
N/A
1 m
10 m
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Length (Metric)
13880710 Remote Dispensing System N/A
13880711 FEP Tubing 10 m
13880712 FEP Tubing 1 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 13880710 Supplier BRANDTECH™ BRAND™ Supplier No. 704261

Accessories to facilitate remote dispensing with Dispensette™ S or Dispensette™ S Organic bottletop dispenser. Dispenser not included

  • 704261- Remote Dispensing System for Dispensette™. Includes wall mount
  • 704267 - 10 meters of filling tube
  • 704269 - 1 meter filling tube

Specifications

Product Type Remote Dispensing System
Autoclavable Not Autoclavable
Color Translucent/White
Material PTFE, FEP, Borosilicate Glass, Ceramic, Polypropylene
For Use With (Application) Dispensing from bulk containers
For Use With (Equipment) Dispensette III/Organic/S/S Organic
Includes Tubing, wall mount, 3/4 in. bung adapter
Sterility Non-sterile
Length (Metric) N/A