missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ XLD Agar (Xylose, Lysine, Desoxycholate)

Catalog No. R01980
Change view
Click to view available options
Quantity:
10/Pk.
Format:
12mm x 85mm Monoplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01980 10/Pk. 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01980 Supplier Thermo Scientific™ Supplier No. R01980

Thermo Scientific™ Remel XLD Agar (Xylose, Lysine, Desoxycholate) is for the selective and differential isolation of gram-negative enteric pathogens.

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, USP.
Description XLD Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.