missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Urea-PDA Disk

Catalog No. R21152
Change view
Click to view available options
Quantity:
25 Disks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21152 25 Disks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21152 Supplier Thermo Scientific™ Supplier No. RE21152

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Demonstrate urease activity and phenylalanine deamination in Enterobacteriaceae with this reagent-impregnated disk.

Demonstrate urease activity and phenylalanine deamination in Enterobacteriaceae with the reagent-impregnated Thermo Scientific™ Remel™ Urea-PDA Disk. Urease activity has been shown to differentiate Proteus and Providencia spp. from other organisms1,2. In 1974, Ederer et al. incorporated urea and phenylalanine into one medium for use as a rapid test3. Golden et al. impregnated filter paper strips with urea and phenylalanine to differentiate members of the Enterobacteriaceae4.

Henriksen and Closs demonstrated that Proteus strains could be differentiated from other enteric Gram-negative bacilli by their action on phenylalanine5,6. In 1950, Henriksen demonstrated that the rapid transformation of phenylalanine into phenylpyruvic acid nearly always coincides with strong urease activity and is practically limited to members of the Proteus group7.

  • Easy to use - Rapid test
  • Easy to interpret - Visible color change

The Urea-PDA disk contains urea and phenylalanine. Proteus produces the enzyme urease, which hydrolyzes urea to form ammonia. Ammonia that is produced causes the phenol red indicator to change from clear to dark pink or red. Proteus and Providencia spp. produce a type of amino acid oxidase enzyme, which causes oxidative deamination of phenylalanine yielding phenylpyruvic acid. When 10% ferric chloride is added to this, it chelates with phenylpyruvic acid to form a green color.

Specifications

Content And Storage 2°C to 8°C
Description Urea-PDA Disk is used to determine urease activity and phenylalanine deamination in Enterobacteriaceae
Detectable Analytes Phenylpyruvic Acid
Format Vial
Quantity 25 Disks