missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Tryptic Soy Broth
The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
Supplier: Thermo Scientific™ R112740
Description
Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.
- Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth
Specifications
| Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP. | |
| BB - 1000mL | |
| 4/Pk. |
| Tryptic Soy Broth | |
| Microbiology Media | |
| 4 x 1000 mL |
REF numbers beginning with R11 are not intended for IVD use.