missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Broth

Catalog No. R112758
Change view
Click to view available options
Quantity:
10 x 25 mL
10 x 100 mL
10 x 500 mL
4 x 1000 mL
Format:
BB - 1000mL
BB Septum - 100mL
BB Septum - 500mL
SB - 100mL
SB - 25mL
WMB Lug - 100mL
Packaging Type:
10/Pk.
4/Pk.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format Packaging Type
R112758 10 x 25 mL SB - 25mL 10/Pk.
R112730 10 x 100 mL WMB Lug - 100mL 10/Pk.
R112731 10 x 100 mL SB - 100mL 10/Pk.
R112732 10 x 500 mL BB Septum - 500mL 10/Pk.
R112740 4 x 1000 mL BB - 1000mL 4/Pk.
R112745 10 x 100 mL BB Septum - 100mL 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. R112758 Supplier Thermo Scientific™ Supplier No. R112758

The Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.

Thermo Scientific™ Remel Tryptic Soy Broth (TSB) is a general purpose enrichment broth for cultivation and isolation of microorganisms.

  • Part numbers beginning with R11 are for sterility testing according to USP (United States Pharmacopeia) and for use as general purpose enrichment broth

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMWW, IDF, SMD, USDA, USP.
Description Tryptic Soy Broth
Format SB - 25mL
Product Type Microbiology Media
Packaging Type 10/Pk.
Quantity 10 x 25 mL

REF numbers beginning with R11 are not intended for IVD use.