missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Tryptic Soy Broth with 4% Polysorbate 20 and 0.5% Lecithin

Catalog No. R112769
Change view
Click to view available options
Quantity:
10 x 90 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R112769 10 x 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R112769 Supplier Thermo Scientific™ Supplier No. RE112769

Thermo Scientific™ Remel Tryptic Soy Broth with 4% Polysorbate 20 and 0.5% Lecithin is a general-purpose enrichment broth for cultivation/isolation of microorganisms.

Thermo Scientific™ Remel Tryptic Soy Broth with 4% Polysorbate 20 and 0.5% Lecithin is a general-purpose enrichment broth for cultivation an isolation of microorganisms.

Specifications

Description Tryptic Soy Broth with 4% Polysorbate 20 and 0.5% Lecithin
Format WMB Lug - 90mL
Product Type Microbiology Media
Quantity 10 x 90 mL

For Sterility Testing. Not intended for IVD use.