missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ Tryptic Soy Agar
The Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.
Supplier: Thermo Scientific™ R112700
Description
Thermo Scientific™ Remel Tryptic Soy Agar (TSA) (Soybean Casein Digest Agar) is a general purpose medium for cultivation and isolation of microorganisms.
Specifications
| Industrial Reference: AOAC, BAM, COMPF, IDF, USDA, USP, SMWW. | |
| BB - 500mL | |
| 10/Pk. |
| Tryptic Soy Agar | |
| Agar | |
| 10 x 500 mL |
REF R110092, REF R112700, and REF R112703 not intended for IVD use.