missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Tryptic Soy Agar with Lecithin, Polysorbate 80 (Microbial Content Test Agar)

Catalog No. R119841
Change view
Click to view available options
Format:
12mm x 85mm Monoplate
20mm x 150mm Pour Tube
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R119841 20mm x 150mm Pour Tube
R01919 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R119841 Supplier Thermo Scientific™ Supplier No. RE119841

Detect and enumerate microorganisms present on surfaces of sanitary importance using this medium.

Detect and enumerate microorganisms present on surfaces of sanitary importance using Thermo Scientific™ Remel™ Tryptic Soy Agar with Lecithin, Polysorbate 80 (Microbial Content Test Agar). The combination of lecithin and polysorbate 80 in an agar medium effectively neutralizes disinfectants1. Tryptic Soy Agar (TSA) is prepared according to the Soybean-Casein Agar formula recommended in the United States Pharmacopeia (USP)2. TSA with Lecithin and Polysorbate 80 is compatible with environmental air sampling procedures.

Tryptic Soy Agar with Lecithin, Polysorbate 80 (Microbial Content Test Agar) is a solid medium recommended for use in qualitative procedures for the detection and enumeration of microorganisms.

Casein peptone and soy peptone provide essential amino acids, peptides and nitrogenous substances necessary for the growth of bacteria. Sodium chloride is a source of essential electrolytes and maintains osmotic equilibrium. Lecithin and polysorbate 80 neutralize residual compounds that may be present on the material being sampled. Agar is a solidifying agent.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Description Tryptic Soy Agar w/Lecithin, Polysorbate 80
Format 20mm x 150mm Pour Tube
Product Type Agar Plate
Quantity 100 x 22 mL

Part Nos. R112718 and R119841 not intended for IVD use.