Learn More
Thermo Scientific™ Remel™ TB Methylene Blue Counterstain
A counterstain for AFB and Cryptosporidium modified acid-fast stain using TB Kinyoun or TB Ziehl-Neelsen stains.
$308.77 - $308.77
Description
Thermo Scientific™ Remel TB Methylene Blue is a counterstain for AFB and Cryptosporidium modified acid-fast stain using TB Kinyoun or TB Ziehl-Neelsen stains.