missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ TAT Broth

Catalog No. R112612
Change view
Click to view available options
Quantity:
1000 mL
10 x 90 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R112612 1000 mL
R112611 10 x 90 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R112612 Supplier Thermo Scientific™ Supplier No. R112612

Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.

Thermo Scientific™ Remel TAT Broth (w/Polysorbate 20) is used for sterility testing of highly viscous substances, such as salves, ointments and cosmetics.

Specifications

Description TAT Broth, w/Polysorbate 20
Format PB -1000mL
Product Type Broth
Quantity 1000 mL

Not intended for IVD use.