missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SPS Disk

Catalog No. R21140
Change view
Click to view available options
Quantity:
25 Disks/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21140 25 Disks/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21140 Supplier Thermo Scientific™ Supplier No. RE21140

missing translation for 'validMainframeMsg'

SPS Disk is a reagent-impregnated disk recommended for presumptive identification of Peptostreptococcus anaerobius.

Presumptively identify Peptostreptococcus anaerobius using Thermo Scientific™ Remel™ SPS Disk. Graves et al. used a reagent-impregnated disk to determine the susceptibility of anaerobic cocci to SPS. They reported that amongst anaerobic cocci only P. anaerobius is sensitive to SPS. Wideman and Wilkins further verified these findings and reported 98% overall accuracy in the identification of P. anaerobius isolated from clinical specimens1,2.

Peptostreptococcus anaerobius is a slow growing, Gram-positive anaerobic cocci (GPAC) bacterium. P. anaerobius is an opportunistic pathogen that can cause multiple deep organ abscesses and infections throughout the body, especially in the abdominal cavity and female urogenitary tract.

Use SPS Disk for presumptive identification of Peptostreptococcus anaerobius.

  • Ready to use - Reagent impregnated disk
  • Easy to store - No special storage required
  • Easy to interpret - Visible zone of inhibition
    • Sensitive - Zone of inhibition equal to or greater than 12 mm
    • Resistant - No zone of inhibition or zone less than 12 mm

SPS Disk contains sodium polyanethol sulfonate. Sodium polyanethol sulfonate (SPS) is a synthetic, heat stable anticoagulant which inhibits bactericidal agents in blood3. SPS has also been found to inhibit the growth of certain anaerobic cocci. The growth of P. anaerobius is inhibited by SPS. Positive test (sensitive) is indicated by a zone diameter of 12-27 mm around SPS disk.

Specifications

Content And Storage 2°C to 8°C
Description SPS Disk
Detectable Analytes P. anaerobius
Format Reagent Disk
Quantity 25 Disks/Vial