missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ SP4 Glucose Broth

Catalog No. R20057
Change view
Click to view available options
Format:
CSB - 100mL
CSB - 90mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R20057 CSB - 90mL
R112585 CSB - 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R20057 Supplier Thermo Scientific™ Supplier No. RE20057

Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.

Thermo Scientific™ Remel SP4 Glucose Broth (w/thallium acetate, penicillin) is used for isolation of Mycoplasma species.

  • Penicillin inhibits gram-positive bacteria

Specifications

Description SP4 Glucose Broth w/thallium acetate, penicillin
Format CSB - 90mL
Product Type Broth
Quantity 90 mL

REF R112585 not intended for IVD use.