Learn More
Thermo Scientific™ Settling Plate Tryptic Soy Agar
Catalog No. R111570
Change view
Click to view available options
Format:
85mm Plate, Double
Settling Monoplate, Double
Settling Monoplate, Single
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
R111570 Settling Monoplate, Single
R111670 Settling Monoplate, Double
R111890 85mm Plate, Double
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. R111570 Supplier Thermo Scientific™ Supplier No. RE111570
Isolate and cultivate wide variety of organisms like bacteria, yeast and fungi using Settling Plate Tryptic Soy Agar.
Isolate and cultivate wide variety of organisms such as bacteria, yeast and fungi using Thermo Scientific™ Remel™ Settling Plate Tryptic Soy Agar. This medium is listed in Standard Methods for the Examination of Water and Wastewater and the Compendium of Methods for the Microbiological Examination of Foods1,2. This medium is recommended for use in Official Methods of Analysis of AOAC International, the International Dairy Federation (IDF), and the United States Department of Agriculture (USDA)3,4. Tryptic Soy Agar slants may be used to cultivate, store, and ship bacterial cultures.
  • Settling plates (single bag and double bag) are manufactured in a clean room environment
  • This provides the highest level of sterility assurance with no compromise of growth characteristics
  • Each lot is tested to ensure consistent, reproducible results
Specifications
Description Settling Plate Tryptic Soy Agar
Format Settling Monoplate, Single
Product Type Agar Plate
Quantity 10/Pk.
Not intended for IVD use.