Learn More
Thermo Scientific™ Remel™ Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated)
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated) facilitates cultivation of yeast and fungi.
$112.74 - $112.74
Specifications
| Description | Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 |
|---|---|
| Format | Monoplate - Double + Bag |
| Product Type | Agar |
Description
Thermo Scientific™ Remel Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 (Irradiated) facilitates cultivation of yeast and fungi.
- Settling Plates (Single Bag and Double Bag) are manufactured in a clean room environment
- This provides the highest level of sterility assurance with no compromise of growth characteristics
- Each lot is tested to ensure consistent, reproducible results
-
Specifications
| Settling Plate Sterile Sabouraud Dextrose Agar pH 5.6 | |
| Monoplate - Double + Bag | |
| Agar |
Not intended for IVD use.