Learn More
Thermo Scientific™ Settling Plate Sterile R2A
Enumerate and cultivate heterotrophic bacteria from potable water using Settling Plate Sterile R2A.
Supplier: Thermo Scientific™ R111970
Description
Enumerate and cultivate heterotrophic bacteria from potable water using Thermo Scientific™ Remel™ Settling Plate Sterile R2A. The growth of stressed and chlorine-tolerant bacteria recovered from treated waters is stimulated using a less nutritious medium, lower incubation temperatures, and longer incubation times. R2A Agar is recommended by the American Public Health Association (APHA) in Standard Methods for the Examination of Water and Wasterwater1. Information on water quality, such as heterotrophic plate counts, may be used to evaluate the efficiency of treatment processes.
Use R2A Agar for enumeration and cultivation of heterotrophic bacteria from potable water. Reasoner and Geldreich of the U.S. Environmental Protection Agency developed R2A Agar for the isolation and recovery of aerobic and facultative heterotrophic bacteria from treated potable water samples3. In this medium the low nutrients give higher counts in treated potable waters as compared to a highly nutritious medium because these bacteria tend to be slow-growing species and would quickly be suppressed by faster-growing species on a richer culture medium.
- Recommended by the APHA in Standard Methods for the Examination of Water and Wasterwater1
- Superior to plate count agar when using pour plate, spread plate and membrane filter technique2.
- Ready-to-use – Convenience of prepared medium
This medium contains peptone that supplies nitrogenous compounds and amino acids necessary for growth. Dextrose is added as a carbon source of energy. Yeast extract supplies B-complex vitamins and serves as a growth enhancer. Addition of soluble starch as a neutralizer and sodium pyruvate stimulates the growth of stressed bacteria. Buffering agents such as dipotassium phosphate and magnesium sulfate maintain osmotic equilibrium.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
| Settling Plate Sterile R2A | |
| Double Bag | |
| Agar | |
| 10/Pk. |
Not intended for IVD use.