missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Dextrose Broth, 2%

Catalog No. R112554
Change view
Click to view available options
Quantity:
10 x 100 mL
Format:
WMB Lug - 100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R112554 10 x 100 mL WMB Lug - 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R112554 Supplier Thermo Scientific™ Supplier No. R112554

Cultivates pathogenic yeast and fungi from clinical specimens with Sabouraud Dextrose Broth 2%.

Cultivates pathogenic yeast and fungi from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Broth 2%. Sabouraud Dextrose Agar is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concentration of dextrose from 40 to 20g/L2. Sabouraud Dextrose Broth 2% is a modification of the original formulation, without agar.

Cultivate pathogenic yeast and fungi from clinical specimens with Sabouraud Dextrose Broth 2%.

  • Ready-to-use – Convenience of a prepared media.
  • Suitable for isolation of pathogenic fungi from a material containi

Specifications

Certifications/Compliance Conforms to USP.
Description Sabouraud Dextrose Broth 2%
Format WMB Lug - 100mL
Product Type Broth
Quantity 10 x 100 mL