Learn More
Thermo Scientific™ Remel™ Sabouraud Dextrose Agar w/Chloramphenicol
Catalog No. R112552
Change view
Click to view available options
Quantity:
10 x 500 mL
Catalog No. Quantity
R112552 10 x 500 mL
1 options
Catalog No. R112552 Supplier Thermo Scientific™ Supplier No. R112552
Thermo Scientific™ Remel Sabouraud Dextrose Agar w/Chloramphenicol (pH 5.6) is used for cultivation and isolation of yeast and fungi.
Thermo Scientific™ Remel Sabouraud Dextrose Agar w/Chloramphenicol (pH 5.6) is used for cultivation and isolation of yeast and fungi.
  • Low pH 5.6 enhances growth of fungi, especially dermatophytes and is slightly inhibitory to bacteria
  • Chloramphenicol inhibits some bacteria
Specifications
Description Sabouraud Dextrose Agar w/Chloramphenicol, pH 5.6
Format BB - 500mL
Quantity 10 x 500 mL
Not intended for IVD use.