missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sabouraud Dextrose Agar (Emmons)

Catalog No. R064422
Change view
Click to view available options
Quantity:
10/Pk.
20/Pk.
Format:
15mm x 103mm Tube
Monoplate Deep Pour
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R064422 20/Pk. 15mm x 103mm Tube
R01760 10/Pk. Monoplate Deep Pour
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R064422 Supplier Thermo Scientific™ Supplier No. RE064422

Isolate and cultivate pathogenic yeast and molds from clinical specimens with this selective medium.

Isolate and cultivate pathogenic yeast and molds from clinical specimens with Thermo Scientific™ Remel™ Sabouraud Dextrose Agar (Emmons) prepared media plates. Sabouraud Dextrose Agar (Emmons) is a modification of the original medium developed by Sabouraud in 18921. Emmons neutralized the pH, increasing it to 7.0, and reduced the concertation of dextrose from 40 to 20 g/L2.

Use Sabouraud Dextrose Agar (Emmons) for primary isolation and cultivation of pathogenic yeasts and molds from clinical specimens.

  • Suitable for isolation of pathogenic fungi from a material containing a large amount of other fungi or bacteria.
  • Ready prepared convenience of prepared media.

Casein and meat peptones supply nitrogenous compounds and amino acids necessary for the growth of fungi. Dextrose is an energy source which serves to support the growth of fungi. Chloramphenicol and gentamicin are broad-spectrum antibiotics which inhibit a range of Gram-positive and Gram-negative bacteria. Cycloheximide is inhibitory to rapidly growing saprophytic fungi that could overgrow slow-growing primary pathogens. Penicillin is active against Gram-positive bacteria. Streptomycin inhibits Gram-positive and Gram-negative bacteria.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Description Sabouraud Dextrose Agar, Emmons
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20/Pk.

REF R112555 not intended for IVD use.