missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Rappaport-Vassiliadis Salmonella Enrichment Broth MLT

Catalog No. R117661
Change view
Click to view available options
Quantity:
20 x 10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R117661 20 x 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R117661 Supplier Thermo Scientific™ Supplier No. R117661

Thermo Scientific™ Remel Rappaport-Vassiliadis Salmonella Enrichment Broth MLT facilitates selective enrichment of Salmonella spp.

Thermo Scientific™ Remel Rappaport-Vassiliadis Salmonella Enrichment Broth MLT facilitates selective enrichment of Salmonella spp.

Specifications

Certifications/Compliance Industrial Reference: USP.
Description Rappaport-Vassiliadis Salmonella Enrichment Broth MLT
Format 16mm x 125mm Tube
Product Type Microbiology Media
Quantity 20 x 10 mL

Not intended for IVD use.