Learn More
Thermo Scientific™ Remel™ QC-Slide™ Gram Stain Control
Catalog No. R40140
Change view
Click to view available options
Quantity:
10 Slides/Pk.
45 Slide/Pack
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R40140 10 Slides/Pk.
R40142 45 Slide/Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R40140 Supplier Thermo Scientific™ Supplier No. R40140
Remel QC-Slide Gram Stain Control is a heat-fixed quality control slide used for Gram stain procedures.
Remel QC-Slide Gram Stain Control is a heat-fixed quality control slide used for Gram stain procedures.
Specifications
Description Gram Stain Control
Format Glass Slide
Product Type Gram Stain Control
Packaging Type 10 Slide/Pack
Quantity 10 Slides/Pk.