missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Potassium Hydroxide 3%

Catalog No. R21251
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21251 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21251 Supplier Thermo Scientific™ Supplier No. NRE0538

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Differentiates gram-positive and gram-negative microorganisms with Potassium Hydroxide 3%.

Differentiate gram-positive and gram-negative organisms with Thermo Scientific™ Remel™ Potassium Hydroxide 3%.Halebian et al., in 1981 reported the use of 3% Potassium Hydroxide (KOH) to differentiate anaerobic gram-positive and gram-negative bacteria1. Von Graevenitz and Bucher further investigated this with other bacteria2.

Potassium Hydroxide 3% is used to differentiate gram-positive and gram-negative microorganisms. Gram-negative cell walls are broken down by 3% KOH and it releases viscoid chromosomal material which causes suspension to become thick and stringy. This reaction characterizes the gram-negative bacteria.Ready to use: Convenience of prepared reagentEasy to interpret: Stringy, viscous material pulled up from suspension within first 30 seconds is easy to interpret.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 30°C
Description Ea.
Includes gram-positive and gram-negative microorganisms.
Quantity 25 mL
Form Liquid