missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Potassium Hydroxide 10%

Catalog No. R21230
Change view
Click to view available options
Quantity:
25 mL/Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21230 25 mL/Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21230 Supplier Thermo Scientific™ Supplier No. RE21230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Directly detect fungal elements in clinical specimens by microscopic examination using 10% Potassium Hydroxide preparation.

Detect fungal elements in clinical specimens by microscopic examination using Thermo Scientific™ Remel™ 10% Potassium Hydroxide preparation. The Laboratory Handbook of Medical Mycology recommends the use of 10% KOH for direct detection of fungal elements1.The KOH preparation, when positive, provides valuable clues as to the etiological agent of infection. McGowan reported the use of 10% KOH but warned that a lack of contrast between fungal elements and clinical material may occur2. Hence, for improved visibility phase contrast microscopy is recommended.

Directly detect fungal elements in clinical specimens by microscopic examination using 10% Potassium Hydroxide preparation. KOH is a strong base.Ready for Use: Convenience of a prepared reagent.Quick and inexpensive detection of fungal elements in clinical specimensKOH acts to disrupt cellular sheets or clumps of proteinaceous debris that may be present and clears the specimen for easier fungal detection3. The addition of glycerol to the KOH solution reduces the amount of precipitation that forms in the reagent containers and prevents KOH preparations from drying4.Not all products are available for sale in all territories. Please inquire.

Specifications

Content And Storage 2°C to 30°C
Description Potassium Hydroxide 10%
Detectable Analytes Fungus
Format Bottle
Quantity 25 mL/Bottle