missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Polysorbate 80 Hydrolysis Substrate Concentrate

Catalog No. R21276
Change view
Click to view available options
Quantity:
1 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21276 1 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21276 Supplier Thermo Scientific™ Supplier No. RE21276

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Differentiate Mycobacterium spp., particularly the slow growing scotochromogens and nonphotochromogens, using Polysorbate 80 Hydrolysis Substrate Concentrate reagent.

Differentiate Mycobacterium spp. particularly the slow growing scotochromogens and nonphotochromogens based on lipase production and the hydrolysis of Polysorbate 80 using Thermo Scientific™ Remel™ Polysorbate 80 Hydrolysis Substrate Concentrate. In 1962, Wayne introduced the test for detecting polysorbate 80 hydrolysis1. In 1973, Kilburn et al. developed a more stable polysorbate hydrolysis test2,3.

Nonpathogenic, slow growing scotochromogens and nonphotochromogens commonly produce lipase which hydrolyzes polysorbate 80 into oleic acid and polyoxyethylated sorbitol, whereas pathogenic Mycobacterium spp. do not produce this enzyme4. Polysorbate 80 Hydrolysis Substrate Concentrate contains the indicator, neutral red dye. Neutral red dye is normally red at pH 7.0, but after binding with polysorbate 80 it changes to amber or straw color that it has at an alkaline pH. Certain Mycobacterium spp. hydrolyse polysorbate 80 which is then no longer available to bind the indicator causing the neutral red dye to revert to its original red color5,6.This reagent contains neutral red as an indicator, disodium and monopotassium phosphate buffers and Polysorbate 80.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description Polysorbate 80 Hydrolysis Substrate Concentrate
Includes 5mL/Tube
Quantity 1 x 5 mL
Form Liquid