missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Polysorbate 80, 10%

Catalog No. R21275
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21275 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21275 Supplier Thermo Scientific™ Supplier No. RE21275

Differentiate and identify catalase producing Mycobacterium spp. using Polysorbate 80, 10% reagent.

Differentiate and identify Mycobacterium spp. with the catalase test using Thermo Scientific™ Remel™ Polysorbate 80, 10% reagent. Most strains of mycobacteria produce catalase1. The catalase test is a semiqualitative test which was developed to help in differentiation of organisms included in Mycobacterium tuberculosis complex (drug-resistant strains) from other Mycobacterium spp2 (non-drug-resistant strains)3,4.

Polysorbate 80 helps in dispersing the hydrophobic, tightly clumped mycobacterial cells from large aggregates to individual bacilli, thus maximizing the detection of catalase. 10% Polysorbate 80 permits more rapid entry of compounds, such as 30% hydrogen peroxide into the mycobacterial cell by lowering the surface tension in a medium.Ready-to-use: Liquid reagentEasy to interpret:Strong Positive Test: A column of bubbles > 45mmWeak Positive Test: A column of bubbles < 45mmNegative Test: No bubblesThis reagent contains 0.2 g/1000mL of Polysorbate 80 (CAS 9005-65-6) supplied in 10mL bottles.Not all products are available for sale in all territories. Please inquire.Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description Polysorbate 80, 10%
Format Vial
Quantity 10 mL