missing translation for 'onlineSavingsMsg'
Learn More

Remel™ Buffered Peptone Water BR 400 mL

Catalog No. R112133
Catalog No. R112133 Supplier Thermo Fisher Scientific Supplier No. R112133

Our Bird rinse bottles are designed to meet the industry's needs for rinsing and collecting poultry carcass rinse samples for pathogen testing.

Introducing our new Bird Rinse Bottles, specifically designed to address the industry's needs for rinsing and collecting poultry carcass rinse samples for further pathogen testing.

These bottles offer improved ergonomics with a modern design, ensuring ease of use. Additionally, they are stackable, enhancing safety during transportation and storage.

With our range of solutions and the added benefits of improved ergonomics, stackable bottles, and reduced contamination risk with no foil rip-off step, we are confident that our Bird Rinse Bottles will meet your needs for rinsing and collecting poultry carcass rinse samples for further pathogen testing.

R112133: Remel™ Buffered Peptone Water BR 400 mL, 24/pk

Buffered Peptone Water: effective for detecting microorganisms on poultry carcasses but may result in lower detection rates of Salmonella if sanitizer residues are carried over into the rinse fluid.

Specifications

Content And Storage 2° to 25°C
Description Remel Buffered Peptone Water BR 400 mL
Quantity 24/pk