missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ PHA Purified

Catalog No. R30852801
Change view
Click to view available options
Quantity:
1 Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R30852801 1 Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R30852801 Supplier Thermo Scientific™ Supplier No. R30852801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Phytohaemagglutinin (PHA) is a mitogenic agent which is used in studies involving cell permeability.

Carry out studies involving cell permeability with Thermo Scientific™ Remel™ Phytohaemagglutinin (PHA). PHA is a mitogenic agent derived from extracts of Phaseolus vulgaris seeds and has twin properties of causing erythroagglutination and of stimulating progressive lymphocyte mitosis in cell culture. PHA has proved to be beneficial in the study of the immune response, lymphocyte kinetics, and bone marrow dynamics, and for these purposes it is desirable to use a substance which has more closely reproducible qualities and known potency.

Purified phytohaemagglutinin is a freeze-dried, highly refined protein fraction of selected Phaseolus spp. seed extract in which the specific mitogenic activity and the mitogenic/ haemagglutinating activity ratio have been increased by a factor of about 100/1 during purification. Both agglutinating and mitogenic activities of PHA are found to be associated with the protein fractions of crude extracts, which are sufficiently alike in their physico-chemical properties, thus it is difficult to separate them. Relatively crude extracts selected for their ability to yield good mitotic pictures have proved to be useful in routine lymphocyte culture for chromosome studies.

  • Ready to use - Prepared reagent
  • Easy to store - Store at 2° to 8°C until use
  • Can be used in routine lymphocyte culture for chromosome studies

Purified phytohaemagglutinin contains a freeze-dried, highly refined protein fraction of selected Phaseolus spp. seed extract. Each bottle contains 2mg lyophilised phytohaemagglutinin (purified), dried from a small volume of buffered saline without preservative. The reagent may be reconstituted to any desired volume in sterile saline (if buffered, preferably to a pH between 6.5 and 8) or distilled water, using a sterile disposable hypodermic syringe.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description PHA Purified
Format Vial
Quantity 1 Vial