missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Pathotec™ and CO Paper Strips

Catalog No. R38191
Change view
Click to view available options
Quantity:
4 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R38191 4 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R38191 Supplier Thermo Scientific™ Supplier No. RE38191

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ Remel Pathotec and CO are reagent impregnated paper strips used for the detection of cytochrome oxidase enzyme.

Thermo Scientific™ Remel Pathotec and CO are reagent impregnated paper strips used for the detection of cytochrome oxidase enzyme.

Specifications

Content And Storage 2°C to 8°C
Description Pathotec™ CO
CE Marker Yes
Format Vial, Strip
Packaging Type 4 Vials/Pk.
Quantity 4 Vials