Learn More
Thermo Scientific™ Remel™ PathoDxtra™ Strep Grouping Kit
Eliminates need for incubation and provides a faster strep grouping solution than more traditional enzyme extraction.
Supplier: Thermo Scientific™ DR0707G
Description
Eliminate the need for incubation and provide a faster strep grouping solution than more traditional enzyme extraction with the Thermo Scientific™ PathoDxtra™ Strep Grouping Kit.
- Improves workflow efficiencies — Instant acid extraction without the need for incubation or reconstitution
- Saves time — Simple, rapid procedure produces easy-to-read results
- Reduces costs — Identifies all clinically important Strep groups, including Group D; no additional reagents required
- Provides flexibility — Pack variants available to suit testing needs
Includes:
PathoDxtra Strep Grouping Kit (Mfr. No. DR0700M): 3mL each of A, B, C, D, F and G grouping latex; Polyvalent Positive Control, 2.8mL; Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; Mixing Sticks; Disposable Reaction Cards
PathoDxtra Extraction Reagent Set (Mfr. No. DR0709M): Extraction Reagent 1: 4mL; Extraction Reagent 2: 4mL; Extraction Reagent 3: 2 x 10mL; For use with the PathoDxtra Strep Grouping Kit
PathoDxtra Strep Grouping Reagent Set (Mfr. No. DR0710M): Extraction Reagent 1 - 4mL; Extraction Reagent 2 - 4mL; Extraction Reagent 3 - 2 x 10mL; Mixing Sticks, Disposable Reaction Cards; For use with selected, separately purchased PathoDxtra Strep Grouping latex reagents
Spécifications
| Positive | |
| beta-haemolytic streptococci | |
| Enzyme Immunoassay Detection | |
| Group A, B, C, D, F and G |
| Polyvalent Positive Control | |
| Positive Control | |
| Polyvalent | |
| 2.8 mL |
Rapid Streptococcal grouping tests correlate best with reference methods when only Streptococci that are Β-hemolytic on sheep blood agar are tested. 1, 2, 3, 4