missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Remel™ PathoDx™ Extraction Reagent 3
Description
Remel PathoDx and Extraction Reagent 3 is used with Strep A Latex Agglutination Kit and Strep Grouping Kit.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | PathoDx Extraction Reagent 3 |
| CE Marker | Yes |
| Quantity | Each |