Learn More
Thermo Scientific™ Remel™ Oxy-Swab™
Catalog No. R249348
Change view
Click to view available options
Quantity:
4 x 25 Swabs
Catalog No. Quantity
R249348 4 x 25 Swabs
1 options
Catalog No. R249348 Supplier Thermo Scientific™ Supplier No. R249348
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
For detection of cytochrome oxidase in oxidase-producing microorganisms, such as Neisseria spp. and Pseudomonas spp.
Thermo Scientific™ Remel Oxy-Swab is used for detection of cytochrome oxidase in oxidase-producing microorganisms, such as Neisseria spp. and Pseudomonas spp.
Ordering Alerts:
Packaged in resealable foil envelopes. 25 Swabs/Envelope.
Specifications
Content And Storage 2°C to 8°C
Description Detects cytochrome oxidase in oxidase-producing microorganisms, such as Neisseria spp. and Pseudomonas spp.
Detectable Analytes Cytochrome Oxidase
Packaging Type 4 Env/Pk.
Product Line Remel, Oxy-Swab
Quantity 4 x 25 Swabs