missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nutrient Agar

Catalog No. R01640
Change view
Click to view available options
Quantity:
10/Pk.
Format:
12mm x 85mm Monoplate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R01640 10/Pk. 12mm x 85mm Monoplate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01640 Supplier Thermo Scientific™ Supplier No. RE01640
Only null left

Cultivate and maintain wide variety of microorganisms with Nutrient Agar.

Cultivate and maintain wide variety of microorganisms with Thermo Scientific™ Remel™ Nutrient Agar. This is a non-selective medium which was published by APHA in 19171, as a standardized medium for the use in the examination of water, wastewater, dairy products and various foods. This medium is also used for the recovery of non-fastidious microorganisms and to cultivate and maintain stock cultures2,3.

Nutrient Agar is a general purpose culture medium used to cultivate and maintain wide variety of microorganisms.

  • Ready to use
  • General-purpose culture medium Can be used to subculture organisms for ma

Specifications

Certifications/Compliance Industrial Reference: AOAC, BAM, COMPF, SMD.
Description Nutrient Agar
Format 12mm x 85mm Monoplate
Product Type Agar Plate
Quantity 10/Pk.