missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nitrate Disk

Catalog No. R21093
Change view
Click to view available options
Quantity:
25 Disks/Vial
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R21093 25 Disks/Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R21093 Supplier Thermo Scientific™ Supplier No. RE21093

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Determine nitrate reduction by anaerobic bacteria using Nitrate Disks.

Determine nitrate reduction by anaerobic bacteria using Thermo Scientific™ Remel™ Nitrate Disks. In 1977, Widemann et al. utilized a nitrate disk for the detection of nitrate reductase production1. They reported it to be 89% comparable to the more conventional and time-consuming indole nitrate medium assay. Filter paper disks impregnated with potassium nitrate are recommended for presumptive testing of anaerobes2,3.

Determine nitrate reduction by anaerobic bacteria using Nitrate Disks.

  • Ready to use: Reagent-impregnated disk.
  • Rapid test: Results are obtained within 3-5 minutes.
  • Easy to identify: A positive reaction showcases a development of red color after addition of Anaerobic Nitrate Reagent A and B and no color change after addition of zinc dust while in a negative reaction there is no color change post addition of reagent but changes to red after addition of zinc dust.

Organisms that possess nitrate reductase can reduce nitrate to nitrite. Nitrate serves as a source of nitrogen for many bacteria. Molybdate is reported to stimulate synthesis of nitrate reductase4. The first step to nitrate utilization is reduction to nitrite by the removal of one oxygen molecule. Nitrite combines with sulfanilic acid and amino-2-naphthaline to form a red colored end product. Certain organisms can further reduce nitrite to nitrogen, yielding a colorless result. An additional test for the presence of unreacted nitrate must be performed to validate the colorless result. Zinc dust catalyzes the reaction of nitrate to nitrite. Therefore, when zinc dust is added, a red color indicates the presence of unreacted nitrate.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Content And Storage 2°C to 8°C
Description Nitrate disc
Detectable Analytes Nitrite, Free Nitrogen Gas
Format Vial
Quantity 25 Disks/Vial