missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide

Catalog No. R08600
Change view
Click to view available options
Quantity:
100/Pk.
Format:
20mm x 113mm Tube
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
R08600 100/Pk. 20mm x 113mm Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R08600 Supplier Thermo Scientific™ Supplier No. NRE2925

Isolate and cultivate pathogenic fungi from clinical specimens which may be contaminated with bacteria and fungi with this selective medium.

Isolate and cultivate pathogenic fungi, especially from clinical specimens which may be contaminated with bacteria and saprophytic fungi with Thermo Scientific™ Remel™ Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide. Leach, Ford and Whiffen described the use of cycloheximide as a selective agent for the inhibition of saprophytic fungi1. Researchers later found that the addition of both cycloheximide and chloramphenicol achieved more complete selectivity against growth of saprophytic fungi and bacteria2.

The Mycobiotic Agar (Mycocel Agar) with Chloramphenicol and Cycloheximide is recommended for recovery of dimorphic fungi and dermatophytes potentially contaminated with bacteria and saprophytic fungi3,4.

  • Selective: chloramphenicol and cycloheximide in the medium act as selective agents. Cycloheximide inhibits rapidly growing saprophytic fungi that may overgrow slower growing pathogens. Chloramphenicaol is a broad-spectrum antibiotic which inhibits Gram-positive and Gram-negative bacteria as well as Nacardia.
  • Ready prepared: convenience of prepared media.

Not all products are available for sale in all territories. Please inquire.
 

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

.

Specifications

Description Mycobiotic Agar (Mycocel Agar) w/chloramphenicol, cycloheximide
Format 20mm x 113mm Tube
Product Type Agar Plate
Quantity 100/Pk.