missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ Mueller Hinton Broth w/Cations, LHB

Catalog No. R112478
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R112478 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R112478 Supplier Thermo Scientific™ Supplier No. RE112478

Thermo Scientific™ Remel Mueller Hinton Broth w/Cations, LHB (laked horse blood) (Ca, Mg) is used for isolation and/or differentiation of microorganisms or specimens.

Thermo Scientific™ Remel Mueller Hinton Broth w/Cations, LHB (laked horse blood) (Ca, Mg) is used for the isolation and/or differentiation of microorganisms or specimens.

Specifications

Description Mueller Hinton Broth w/Cations, LHB
Format CSB - 100mL
Product Type Microbiology Media
Quantity 100 mL