missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Motility B Medium w/TTC

Catalog No. R061394
Change view
Click to view available options
Quantity:
20 x 5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R061394 20 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R061394 Supplier Thermo Scientific™ Supplier No. RE061394

Demonstrate motility, primarily in nonfermentative, gram negative rods and enterococci, using Motility B Medium w/TTC Indicator.

Demonstrate motility in nonfermentative, gram negative bacilli with Thermo Scientific™ Remel™ Motility B Medium w/TTC Indicator.

Motility B Medium was formulated by Gilardi with a reduced agar concentration to facilitate the appearance of motility by non-fermentative, gram-negative bacilli such as pseudomonads 1,2. The Gard plate technique that contains Motility B Medium poured into a petri dish 3 is used to select a motile strain. Kelly and Fulton recommend the addition of 2,3,5-triphenyltetrazolium chloride (TTC) to enhance the macroscopic appearance of motile organisms in motility media4.

Motility B Medium w/TTC Indicator is a semisolid media that is used to demonstrate motility, primarily in nonfermentative, gram negative bacilli. Agar provides a semisolid medium which makes motility interpretations macroscopic. TTC indicator helps to interpret results. Tetrazolium salt is colorless, but as the organism grows the dye is incorporated into the bacterial cells and is reduced to an insoluble red pigment, formazan. This red color forms only in the area where bacteria is growing.

  • Ready-to-use: Convenience of prepared media.
  • Incubation at room temperature2
  • Easy to interpret:
    • Positive test- In presence of TTC, red color is observed in the area of growth.
    • Negative test- Growth confined to the stab line. No red color.

Motility B Medium w/TTC Indicator contains casein peptone and yeast extract that supply amino acids, peptides and vitamins to promote growth of nonfermentative gram-negative bacilli. Sodium chloride provide essential electrolytes and maintains osmotic equilibrium. 2,3,5-triphenyltetrazolium chloride (TTC) acts as indicator and helps to interpret results by changing color.

 

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Description Motility B Medium w/TTC Indicator
Format 15mm x 103mm Tube
Product Type Microbiology Media
Quantity 20 x 5 mL