Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Supplier: Thermo Scientific™ R20421
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
- Tube size is 15 x 103mm and fits in most spectrophotometers
- Polystyrene technology allows longer shelf life
Includes:
1 each of 0.5, 1.0, 2.0, 3.0, 4.0.
Specifications
| Turbidity Standard Set (1 each of 0.5, 1.0, 2.0, 3.0, 4.0) | |
| 1 each of 0.5, 1.0, 2.0, 3.0, 4.0 | |
| 15 x 103 mm |