Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Supplier: Thermo Scientific™ R20415
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
- Tube size is 15 x 103mm and fits in most spectrophotometers
- Polystyrene technology allows longer shelf life
Chemical Identifiers
| 15 x 103 mm |
Specifications
| Turbidity Standard 5.0 | |
| 15 x 103 mm |