Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Supplier:  Thermo Scientific™ R20413
 View more versions of this product
Catalog No. R20413
Only null left
Description
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
Specifications
Chemical Identifiers
15 x 103 mm
Specifications
Turbidity Standard 3.0
15 x 103 mm
Product Suggestions
Product Suggestions