Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Catalog No. R20412
Change view
Click to view available options
Description:
Turbidity Standard 0.5
Turbidity Standard 1.0
Turbidity Standard 2.0
Turbidity Standard 3.0
Turbidity Standard 4.0
Turbidity Standard 5.0
Turbidity Standard Set (1 each of 0.5, 1.0, 2.0, 3.0, 4.0)
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
R20412 Turbidity Standard 2.0
R20421 Turbidity Standard Set (1 each of 0.5, 1.0, 2.0, 3.0, 4.0)
R20414 Turbidity Standard 4.0
R20413 Turbidity Standard 3.0
R20415 Turbidity Standard 5.0
R20410 Turbidity Standard 0.5
R20411 Turbidity Standard 1.0
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. R20412 Supplier Thermo Scientific™ Supplier No. R20412
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
Specifications
Description Turbidity Standard 2.0
Tubing Size 15 x 103 mm