missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mannitol Salt Agar (Dehydrated)

Catalog No. R453902
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453902 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R453902 Supplier Thermo Scientific™ Supplier No. RE453902

Selectively and differentially isolate staphylococci from clinical and non-clinical samples using Mannitol Salt Agar.

Selectively and differentially isolate staphylococci using Thermo Scientific™ Remel™ Mannitol Salt Agar (Dehydrated). This phenol red mannitol agar contains 7.5% sodium chloride as a selective agent as reported by Koch in 19421. Most strains of coagulase-positive staphylococci grow on this agar, producing yellow zones because of mannitol fermentation, whereas coagulase-negative staphylococci strains produce small colonies with no color change in the surrounding medium. This agar is formulated in conformance with harmonized United States Pharmacopeia (USP)/European Pharmacopeia (EP) guidelines2,3.

Mannitol Salt Agar is a selective and differential solid media for isolating staphylococci by their ability to ferment mannitol.

  • Conforms to USP and EP: Formulated in conformance with harmonized United States Pharmacopeia (USP)/European Pharmacopeia (EP) guidelines2,,3.
  • Easy to differentiate: Differentiates between coagulase-positive and coagulase-negative staphylococci on the basis of mannitol fermentation
  • Selective: Sodium chloride at a concentration of 7.5% inhibits many bacteria other than staphylococci.

This medium contains casein and meat peptones that supply nitrogen, amino acids, and peptides necessary for bacterial growth. Sodium chloride in the concentration of 7.5% is a selective agent that inhibits many bacteria other than staphylococci. Phenol red is the pH indicator which causes a color change in the medium from red-orange to yellow when acid is produced. Colonies of staphylococci that ferment mannitol will be surrounded by a yellow zone, while those that do not ferment mannitol will have no color change (red).

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

Specifications

Certifications/Compliance Industrial Reference: USP
Description Mannitol Salt Agar
Form Powder
Product Type Mannitol Salt Agar
Quantity 500 g

For Laboratory Use Only