missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mannitol Salt Agar

Catalog No. p-4523458
Change view
Click to view available options
Quantity:
10/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R01580 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R01580 Supplier Thermo Scientific™ Supplier No. R01580
Only null left

Selectively and differentially isolate and identify staphylococci from clinical and non-clinical samples using Mannitol Salt Agar.

Selectively and differentially isolate and identify staphylococci from clinical and non-clinical samples using Thermo Scientific™ Remel™ Mannitol Salt Agar. This phenol red mannitol agar contains 7.5% sodium chloride as a selective agent as reported by Koch in 19421. Most strains of coagulase-positive staphylococci grow on this agar, producing yellow zones because of mannitol fermentation whereas coagulase-negative staphylococci strains produce small colonies with no color change in the surrounding medium.

Staphylococci are widespread in nature, although they are mainly found on the skin, skin glands and mucous membranes of mammals and birds. The coagulase-positive species i.e., Staphylococcus aureus is well documented as a human opportunistic pathogen. The ability to clot plasma continues to be the most widely used and accepted criterion for the identification of pathogenic staphylococci associated with acute infections2.

Mannitol Salt Agar is a selective and differential solid media for isolating staphylococci bacteria by using their capability to ferment mannitol by the production of coagulase enzyme.

  • Easy to differentiate: Differentiates between coagulase-positive and coagulase-negative staphylococci.
  • Selective: Sodium chloride in the concentration of 7.5% inhibits many bacteria other than staphylococci.

This medium contains peptones and beef extract, which supply nitrogen, vitamins, minerals, and amino acids essential for growth. The 7.5% concentration of sodium chloride results in the partial or complete inhibition of bacterial organisms other than staphylococci. Sodium chloride also supplies essential electrolytes for transport and osmotic balance. Mannitol is the fermentable carbohydrate, fermentation of which leads to acid production, detected by phenol red indicator, and aids in the differentiation of staphylococcal species. Coagulase positive staphylococci produce yellow colonies and a surrounding yellow medium while coagulase negative staphylococci produce red colonies and no color change of the phenol red indicator. Agar is the solidifying agent.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications

Certifications/Compliance Industrial Reference: BAM
Description Mannitol Salt Agar
Format 12mm x 85mm Monoplate
Product Type Agar
Quantity 10/Pk.