missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MacConkey Broth
Description
Selectively and differentially isolate enteric gram-negative bacilli using MacConkey Broth.
Selectively and differentially isolate enteric gram-negative bacilli using Thermo Scientific™ Remel™ MacConkey Broth. MacConkey Broth is a modification of the original bile salt broth first described by MacConkey in 1900, which contained litmus as an indicator and sodium taurocholate to inhibit gram-positive organisms1. In further testing, MacConkey recommended replacing litmus with neutral red2. In 1953, Childs and Allen replaced neutral red with bromcresol purple, a less inhibitory indicator3.
Also used in broth method for ProSpecT Microplate Assay, Part No. R2474048 and Part No. R2474096.
Specifications
Specifications
| Certifications/Compliance | USP |
| Description | MacConkey Broth |
| Format | PB - 100mL |
| Product Type | Broth |
| Quantity | 10 x 100 mL |