missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Remel™ MacConkey Agar

Catalog No. r453804
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R453804 2.5 kg
R453802 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R453804 Supplier Thermo Scientific™ Supplier No. RE453804
Only null left

The Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.

Thermo Scientific™ Remel MacConkey Agar is used for selective and differential isolation of gram-negative bacilli on the basis of lactose fermentation.

Specifications

Certifications/Compliance Industrial Reference: BAM, COMPF, SMWW, USP.
Description MacConkey Agar
Form Powder
Product Type Agar
Quantity 2.5 kg